Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam24a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183272.2 | NP_899095.1 | 3 | 264 | ||
GenBank mRNA | AK006815.1 | - | 3 | 265 | ||
AK018853.1 | - | 3 | 266 | |||
AK018873.1 | - | 3 | 304 | |||
AK018966.1 | - | 3 | 304 | |||
BC048642.1 | - | 3 | 321 | 6772199 |