Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI846148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033139.3 | NP_001028311.2 | 4 | 904 | ||
NM_001167831.1 | NP_001161303.1 | 4 | 904 | |||
GenBank mRNA | AK090385.1 | - | 4 | 873 | ||
AK142757.1 | - | 3 | 300 | |||
AK163590.1 | - | 4 | 705 | |||
AK166227.1 | - | 4 | 874 | |||
AK168010.1 | - | 4 | 800 | |||
AK212840.1 | - | 2 | 99 | |||
BC125249.1 | - | 4 | 732 | 40059530 |