Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Babam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026636.2 | NP_080912.2 | 8 | 953 | ||
GenBank mRNA | AK077402.1 | - | 8 | 939 | ||
AK147083.1 | - | 8 | 948 | |||
BC005692.1 | - | 8 | 940 | 3597632 |