Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Babam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026636.2 | NP_080912.2 | 9 | 1065 | ||
GenBank mRNA | AK003366.1 | - | 2 | 86 | ||
AK077402.1 | - | 9 | 1051 | |||
AK078265.1 | - | 2 | 86 | |||
AK147083.1 | - | 9 | 1060 | |||
AK210692.1 | - | 5 | 323 | |||
BC005692.1 | - | 9 | 1052 | 3597632 |