Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177576.1 | NP_001171047.1 | 10 | 1134 | ||
NM_026646.3 | NP_080922.1 | 10 | 1376 | |||
GenBank mRNA | AK010193.1 | - | 10 | 1122 | ||
AK018760.1 | - | 10 | 1159 | |||
AK046874.1 | - | 10 | 1121 | |||
AK048389.1 | - | 10 | 1105 | |||
AK049313.1 | - | 10 | 1105 | |||
AK155276.1 | - | 10 | 1128 | |||
AK158985.1 | - | 1 | 4182 | |||
AK162400.1 | - | 10 | 1087 | |||
BC037949.1 | - | 10 | 1133 | 5367108 | ||
BC050887.1 | - | 9 | 1852 |