Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163718.1 | NP_001157190.1 | 3 | 511 | ||
NM_026676.3 | NP_080952.2 | 3 | 532 | |||
GenBank mRNA | AK002309.1 | - | 3 | 504 | ||
AK003675.1 | - | 1 | 983 | |||
AK158743.1 | - | 3 | 533 | |||
AK169072.1 | - | 2 | 867 | |||
AK170074.1 | - | 3 | 501 | |||
BC027751.1 | - | 3 | 602 | 5011894 | ||
BC038884.1 | - | 1 | 305 | 1516275 | ||
BC089019.1 | - | 3 | 481 | 30551363 | ||
CA979987.1 | - | 3 | 460 |