Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026703.2 | NP_080979.1 | 3 | 366 | ||
GenBank mRNA | AK002744.1 | - | 3 | 366 | ||
AK088463.1 | - | 3 | 412 | |||
BC012416.1 | - | 3 | 364 | 3584066 | ||
BC024673.1 | - | 3 | 363 | 4195454 |