Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178065.4 | NP_835166.3 | 17, 18 | 2731 | ||
GenBank mRNA | AB093230.1 | - | 16, 17 | 2350 | ||
AK077015.1 | - | 16, 17 | 2263 | |||
AK133332.1 | - | 16, 17 | 2267 | |||
AK147878.1 | - | 17, 18 | 2695 | |||
AK154529.1 | - | 17, 18 | 2712 | |||
BC006074.1 | - | 8, 9 | 886 | |||
BC049900.1 | - | 11, 12 | 1421 | |||
BC057351.1 | - | 16, 17 | 2559 | |||
BC060658.1 | - | 16, 17 | 2300 | 30355406 | ||
BC079590.1 | - | 17, 18 | 2685 |