Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178065.4 | NP_835166.3 | 19, 20 | 2989 | ||
GenBank mRNA | AB093230.1 | - | 18, 19 | 2608 | ||
AK077015.1 | - | 18, 19 | 2521 | |||
AK133332.1 | - | 18, 19 | 2525 | |||
AK147491.1 | - | 19, 20 | 5204 | |||
AK147878.1 | - | 19, 20 | 2953 | |||
AK154529.1 | - | 19, 20 | 2970 | |||
BC006074.1 | - | 10, 11 | 1144 | |||
BC049900.1 | - | 13, 14 | 1679 | |||
BC057351.1 | - | 18, 19 | 2815 | |||
BC060658.1 | - | 18, 19 | 2558 | 30355406 | ||
BC079590.1 | - | 19, 20 | 2943 |