Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cthrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026778.2 | NP_081054.1 | 4 | 758 | ||
GenBank mRNA | AK003674.1 | - | 4 | 754 | ||
AK076498.1 | - | 4 | 762 | |||
AK151282.1 | - | 4 | 759 | |||
BC131907.1 | - | 4 | 734 | 40130184 | ||
BC144745.1 | - | 3 | 512 | 9053293 |