Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026947.4 | NP_081223.1 | 3, 4 | 347 | ||
GenBank mRNA | AK007578.1 | - | 3, 4 | 348 | ||
AK131590.1 | - | 3, 4 | 348 | |||
BC014724.1 | - | 3, 4 | 321 | 4219360 | ||
FJ687155.1 | - | 2, 3 | 270 |