Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026947.4 | NP_081223.1 | 4 | 388 | ||
GenBank mRNA | AK007578.1 | - | 4 | 389 | ||
AK131590.1 | - | 4 | 389 | |||
BC014724.1 | - | 4 | 362 | 4219360 |