Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex11g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026951.2 | NP_081227.1 | 4 | 457 | ||
GenBank mRNA | AK007582.1 | - | 4 | 458 | ||
AK007848.1 | - | 3 | 266 | |||
BC024096.1 | - | 4 | 488 | |||
BC062135.1 | - | 4 | 446 | 6510277 |