Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700025G04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197990.3 | NP_932107.1 | 5 | 800 | ||
GenBank mRNA | AI552192.1 | - | 5 | 450 | ||
AK006337.1 | - | 6 | 595 | |||
AK148738.1 | - | 4 | 317 | |||
AK167392.1 | - | 451 | ||||
BC034723.1 | - | 5 | 579 | 3981517 | ||
BC055912.1 | - | 5 | 615 | 3978655 |