Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027081.3 | NP_081357.2 | 19 | 1664 | ||
GenBank mRNA | AK006427.1 | - | 10 | 801 | ||
AK084555.1 | - | 17 | 1690 | |||
AK154949.1 | - | 15 | 2317 | |||
BC017544.1 | - | 11 | 1620 | |||
BC030395.1 | - | 17 | 2336 | |||
BC058958.1 | - | 15 | 1171 | |||
BC080797.1 | - | 15 | 1553 | |||
BC139003.1 | - | 19 | 1638 | 8862025 | ||
BQ176037.1 | - | 2 | 360 |