Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027085.3 | NP_081361.1 | 2, 3 | 192 | ||
GenBank mRNA | AK009020.1 | - | 2, 3 | 194 | ||
BC060967.1 | - | 2, 3 | 197 | 30297308 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC051114.1 | - | 208 | Chr. - - [-] on Build |