Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172154.1 | NP_001165625.1 | 3, 4 | 501 | ||
NM_027109.2 | NP_081385.1 | 3, 4 | 618 | |||
GenBank mRNA | AK009174.1 | - | 3, 4 | 624 | ||
AK150242.1 | - | 3, 4 | 531 | |||
BC023246.1 | - | 3, 4 | 501 | 4039470 | ||
DQ116784.1 | - | 3, 4 | 512 |