Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1f8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027163.4 | NP_081439.1 | 3 | 219 | ||
GenBank mRNA | AK009787.1 | - | 3 | 231 | ||
AY071842.1 | - | 3 | 204 | |||
BC119517.1 | - | 3 | 219 | 40055701 |