Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aarsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144829.1 | NP_659078.1 | 6 | 637 | ||
GenBank mRNA | AK078016.1 | - | 6 | 606 | ||
AK168281.1 | - | 6 | 604 | |||
AK189222.1 | - | 2 | 101 | |||
BC005711.1 | - | 6 | 637 | 3497962 |