Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Degs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171002.1 | NP_001164473.1 | 2 | 763 | ||
NM_027299.5 | NP_081575.2 | 2 | 763 | |||
GenBank mRNA | AF466377.1 | - | 2 | 751 | ||
AK008666.1 | - | 2 | 764 | |||
AK162979.1 | - | 2 | 756 | |||
BC016427.1 | - | 2 | 745 | 4218832 | ||
BC035454.1 | - | 2 | 745 |