Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iyd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027391.3 | NP_081667.1 | 1 | 138 | ||
GenBank mRNA | AK002363.1 | - | 1 | 139 | ||
BC023358.1 | - | 1 | 161 | 5064638 |