Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133726.2 | NP_598487.1 | 8 | 713 | ||
GenBank mRNA | AK013981.1 | - | 8 | 699 | ||
AK146528.1 | - | 8 | 709 | |||
AK147169.1 | - | 736 | ||||
AK150686.1 | - | 684 | ||||
AK151631.1 | - | 684 | ||||
AK152270.1 | - | 8 | 709 | |||
AK154658.1 | - | 702 | ||||
AK159149.1 | - | 693 | ||||
AK168557.1 | - | 3 | 235 | |||
AK176982.1 | - | 2 | 74 | |||
BC003843.1 | - | 8 | 713 | 3495262 | ||
CT010318.1 | - | 8 | 613 |