Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190885.1 | NP_001177814.1 | 8 | 996 | ||
NM_001190886.1 | NP_001177815.1 | 7 | 952 | |||
NM_001290690.1 | NP_001277619.1 | 7 | 1151 | |||
NM_027398.3 | NP_081674.2 | 7 | 963 | |||
GenBank mRNA | AB075041.1 | - | 7 | 593 | ||
AJ278534.1 | - | 6 | 521 | |||
AK014313.1 | - | 7 | 951 | |||
AK045948.1 | - | 8 | 997 | |||
AK046398.1 | - | 8 | 985 | |||
AK081845.1 | - | 7 | 953 | |||
AY050525.1 | - | 8 | 601 | |||
AY050526.1 | - | 7 | 568 | |||
AY171234.1 | - | 7 | 1027 | |||
AY647242.1 | - | 7 | 795 | |||
BC034241.1 | - | 7 | 905 | 4503251 | ||
BU702946.1 | - | 6 | 649 | |||
DQ148493.1 | - | 7 | 905 | |||
DQ148494.1 | - | 7 | 953 | |||
DQ148495.1 | - | 8 | 601 |