Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610034B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027420.4 | NP_081696.1 | 3 | 227 | ||
GenBank mRNA | AK011654.1 | - | 3 | 285 | ||
AK039350.1 | - | 3 | 235 | |||
AK051261.1 | - | 3 | 228 | |||
AK156013.1 | - | 3 | 295 | |||
BC031379.1 | - | 3 | 226 | 5360750 |