Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099275.1 | NP_001092745.1 | 4 | 785 | ||
NM_001291001.1 | NP_001277930.1 | 4 | 802 | |||
GenBank mRNA | AK014352.1 | - | 4 | 654 | ||
BC115969.1 | - | 4 | 785 | 40107624 | ||
GQ292766.1 | - | 4 | 802 |