Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099275.1 | NP_001092745.1 | 5 | 1000 | ||
NM_001291001.1 | NP_001277930.1 | 5 | 1017 | |||
GenBank mRNA | AK014352.1 | - | 5 | 868 | ||
BC115969.1 | - | 5 | 1000 | 40107624 |