Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027631.2 | NP_081907.1 | 1 | 1169 | ||
GenBank mRNA | AK016441.1 | - | 1 | 1176 | ||
AK076636.1 | - | 1 | 1170 | |||
BC049750.1 | - | 1 | 1169 | 6773045 |