Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027649.3 | NP_081925.2 | 3 | 420 | ||
GenBank mRNA | AK005865.1 | - | 3 | 421 | ||
AK016586.1 | - | 3 | 347 | |||
AK078278.1 | - | 342 | ||||
AK165435.1 | - | 2 | 208 | |||
AY336501.1 | - | 3 | 308 |