Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027725.3 | NP_082001.2 | 6 | 519 | ||
GenBank mRNA | AK016201.1 | - | 9 | 809 | ||
AK016977.1 | - | 6 | 520 | |||
BC049704.1 | - | 6 | 519 | |||
BC144994.1 | - | 6 | 496 | |||
BC145995.1 | - | 6 | 519 | 40131298 |