Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a19os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027780.1 | NP_082056.1 | 2 | 422 | ||
GenBank mRNA | AK017337.1 | - | 2 | 423 |