Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pogk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142948.1 | NP_001136420.1 | 5 | 1798 | ||
NM_175170.4 | NP_780379.2 | 6 | 2132 | |||
GenBank mRNA | AK031716.1 | - | 6 | 2086 | ||
AK048067.1 | - | 6 | 2091 | |||
AK048752.1 | - | 3 | 1531 | |||
AK053898.1 | - | 6 | 2109 | |||
AK122520.1 | - | 5 | 2070 | |||
BC062177.1 | - | 1 | 568 | |||
BC072587.1 | - | 1752 | 6829796 |