Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023625.4 | NP_076114.2 | 12 | 1786 | ||
GenBank mRNA | AF217003.1 | - | 12 | 1767 | ||
AK004973.1 | - | 12 | 1789 | |||
AK029813.1 | - | 12 | 1785 | |||
AK030234.1 | - | 12 | 1738 | |||
AK032930.1 | - | 12 | 1782 | |||
AK042089.1 | - | 1782 | ||||
AK043608.1 | - | 12 | 1776 | |||
AK150201.1 | - | 12 | 1775 | |||
AK164220.1 | - | 12 | 1767 | |||
AK170387.1 | - | 12 | 1778 | |||
AK170631.1 | - | 12 | 1766 | |||
AK196986.1 | - | 1 | 82 | |||
BC026395.1 | - | 12 | 1764 | |||
BC038605.1 | - | 12 | 1767 | 4921245 | ||
BC048826.1 | - | 12 | 1773 | 4196604 | ||
BC060095.1 | - | 8 | 4176 |