Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suds3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122666.2 | NP_001116138.1 | 6 | 591 | ||
NM_178622.5 | NP_848737.3 | 6 | 591 | |||
GenBank mRNA | AF469109.1 | - | 6 | 531 | ||
AK010274.1 | - | 6 | 494 | |||
AK043652.1 | - | 6 | 534 | |||
AK045475.1 | - | 6 | 545 | |||
AK138947.1 | - | 6 | 542 | |||
AK150633.1 | - | 6 | 529 | |||
AK188755.1 | - | 11 | ||||
BC040088.1 | - | 4 | 204 | |||
BC055764.1 | - | 6 | 544 | 6402735 | ||
BC062176.1 | - | 6 | 458 | 6514171 |