Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023637.3 | NP_076126.2 | 13 | 1248 | ||
GenBank mRNA | AB029949.1 | - | 13 | 1220 | ||
AK010491.1 | - | 13 | 1226 | |||
AK087606.1 | - | 13 | 1215 | |||
AK140760.1 | - | 13 | 1215 | |||
BC079664.1 | - | 13 | 1225 | 30357834 |