Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136237.1 | NP_001129709.1 | 12 | 1775 | ||
NM_028051.3 | NP_082327.2 | 11 | 1861 | |||
NM_028092.3 | NP_082368.1 | 11 | 1737 | |||
GenBank mRNA | AI413122.1 | - | 1 | 166 | ||
AK007473.1 | - | 11 | 1862 | |||
AK008448.1 | - | 11 | 1738 | |||
BC028990.1 | - | 12 | 1736 | 5041383 |