Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172413.2 | NP_766001.1 | 2 | 637 | ||
GenBank mRNA | AK087971.1 | - | 2 | 584 | ||
AK146057.1 | - | 2 | 616 | |||
AK146956.1 | - | 2 | 679 | |||
AK147015.1 | - | 2 | 679 | |||
BC027363.1 | - | 2 | 601 | 4951566 | ||
BC050056.1 | - | 2 | 599 | 30016070 | ||
BC064814.1 | - | 2 | 633 | 6513822 | ||
BC094890.1 | - | 2 | 637 | 30649398 |