Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028185.2 | NP_082461.1 | 3 | 631 | ||
GenBank mRNA | AF514309.1 | - | 3 | 608 | ||
AK008830.1 | - | 1 | 17 | |||
AK082292.1 | - | 3 | 632 | |||
AK143661.1 | - | 3 | 624 | |||
BC094286.1 | - | 3 | 613 | 5294386 |