Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133346.2 | NP_579924.1 | 2 | 321 | ||
GenBank mRNA | AF398967.1 | - | 2359 | |||
AK049312.1 | - | 2 | 310 | |||
AK088890.1 | - | 2 | 256 | |||
AY665653.1 | - | 2 | 207 | |||
BC019192.1 | - | 2 | 313 | 5035395 | ||
BC085485.1 | - | 2 | 278 |