Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289543.1 | NP_001276472.1 | 7, 8 | 1106 | ||
NM_001289544.1 | NP_001276473.1 | 8, 9 | 1430 | |||
NM_028227.3 | NP_082503.2 | 8, 9 | 1302 | |||
GenBank mRNA | AF321920.1 | - | 8, 9 | 1194 | ||
AF321921.1 | - | 8, 9 | 1431 | |||
AK013885.1 | - | 6, 7 | 795 | |||
AK034212.1 | - | 3, 4 | 235 | |||
AK143718.1 | - | 7, 8 | 1096 | |||
BC038490.1 | - | 5, 6 | 671 | 3599718 | ||
BC062932.1 | - | 8, 9 | 1278 |