Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289543.1 | NP_001276472.1 | 5 | 824 | ||
NM_001289544.1 | NP_001276473.1 | 6 | 1148 | |||
NM_028227.3 | NP_082503.2 | 6 | 1020 | |||
GenBank mRNA | AF321920.1 | - | 6 | 912 | ||
AF321921.1 | - | 6 | 1149 | |||
AK013885.1 | - | 4 | 513 | |||
AK143718.1 | - | 5 | 814 | |||
BC038490.1 | - | 3 | 389 | 3599718 | ||
BC062932.1 | - | 6 | 996 |