Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110017D15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048005.1 | NP_001041470.1 | 4 | 590 | ||
NM_001253777.1 | NP_001240706.1 | 4 | 590 | |||
NM_001253778.1 | NP_001240707.1 | 1 | 197 | |||
NM_001253780.1 | NP_001240709.1 | 1 | 197 | |||
NM_001253788.1 | NP_001240717.1 | 4 | 590 | |||
NM_028624.1 | NP_082900.1 | 1 | 197 | |||
GenBank mRNA | AB231294.1 | - | 4 | 589 | ||
AK003742.1 | - | 1 | 188 | |||
BC049629.1 | - | 1 | 187 | |||
CA463601.1 | - | 1 | 200 | |||
DQ873295.1 | - | 1 | 59 | |||
DQ873296.1 | - | 1 | 59 |