Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285959.1 | NP_001272888.1 | 2 | 152 | ||
NM_001285960.1 | NP_001272889.1 | 2 | 285 | |||
NM_029610.3 | NP_083886.1 | 2 | 536 | |||
GenBank mRNA | AK009135.1 | - | 2 | 539 | ||
AK015083.1 | - | 3 | 344 | |||
AK157931.1 | - | 154 | ||||
BC011339.1 | - | 2 | 326 | 3978191 | ||
BY752674.1 | - | 2 | 154 | |||
DV043140.1 | - | 1 | 310 |