Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289602.1 | NP_001276531.1 | 13 | 2263 | ||
NM_028707.4 | NP_082983.3 | 13 | 2269 | |||
GenBank mRNA | AK018116.1 | - | 8 | 725 | ||
AK045529.1 | - | 13 | 2000 | |||
AK053835.1 | - | 13 | 1917 | |||
AK149027.1 | - | 13 | 1999 | |||
BC056437.1 | - | 9 | 768 | |||
BC062930.1 | - | 12 | 1850 | 6855293 | ||
BC065053.1 | - | 13 | 2037 | 5709231 | ||
BC065055.1 | - | 13 | 2020 | 5720949 | ||
BC066026.1 | - | 13 | 2269 | 5721455 | ||
BC066036.1 | - | 13 | 2026 | 6837266 |