Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023734.3 | NP_076223.3 | 5 | 1061 | ||
GenBank mRNA | AB046537.1 | - | 710 | |||
AI587910.1 | - | 3 | 340 | |||
AK004672.1 | - | 4 | 1240 | |||
AK157854.1 | - | 4 | 1552 | |||
AK158339.1 | - | 4 | 1548 | |||
BC039124.1 | - | 5 | 658 |