Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004444.1 | - | 6, 7 | 826 | ||
AK076500.1 | - | 6, 7 | 813 | |||
AK151245.1 | - | 5, 6 | 632 | |||
AK158058.1 | - | 6, 7 | 888 | |||
AK179782.1 | - | 1, 2 | 127 | |||
AK219306.1 | - | 1, 2 | 116 | |||
BC033921.1 | - | 6, 7 | 798 | 5068809 |