Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931431C16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016496.1 | - | 4 | 590 | ||
AK030185.1 | - | 4 | 621 |
Gene Symbol: Acox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028818.1 | - | 1 | 239 |