Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028923.3 | NP_083199.1 | 12 | 1810 | ||
GenBank mRNA | AK016671.1 | - | 12 | 1792 | ||
AK028183.1 | - | 12 | 1794 | |||
AK133888.1 | - | 12 | 1762 | |||
AK145748.1 | - | 12 | 1786 | |||
AK161015.1 | - | 12 | 1756 | |||
AK161659.1 | - | 12 | 1793 | |||
AK180772.1 | - | 2 | 242 | |||
AK207245.1 | - | 2 | 224 | |||
AK210720.1 | - | 2 | 224 | |||
AK216189.1 | - | 2 | 224 | |||
BC026797.1 | - | 12 | 1790 | 3990668 |