Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028923.3 | NP_083199.1 | 13 | 1938 | ||
GenBank mRNA | AK016671.1 | - | 13 | 1920 | ||
AK028183.1 | - | 13 | 1922 | |||
AK133888.1 | - | 13 | 1890 | |||
AK145748.1 | - | 13 | 1914 | |||
AK161015.1 | - | 13 | 1884 | |||
AK161659.1 | - | 13 | 1921 | |||
AK180772.1 | - | 3 | 370 | |||
AK217642.1 | - | 1 | 77 | |||
BC026797.1 | - | 13 | 1918 | 3990668 |