Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd200r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128132.1 | NP_001121604.1 | 3 | 413 | ||
NM_001128133.1 | NP_001121605.1 | 3 | 413 | |||
NM_027578.1 | NP_081854.1 | 3 | 413 | |||
NM_029018.4 | NP_083294.2 | 3 | 413 | |||
GenBank mRNA | AK014637.1 | - | 3 | 387 | ||
AK014671.1 | - | 3 | 350 | |||
AK040868.1 | - | 3 | 400 | |||
AK170363.1 | - | 3 | 343 | |||
AY230199.1 | - | 3 | 387 | |||
AY703837.1 | - | 3 | 318 | |||
AY703838.1 | - | 3 | 318 | |||
AY703839.1 | - | 3 | 318 | |||
AY703840.1 | - | 3 | 318 | |||
AY703841.1 | - | 3 | 318 | |||
AY703842.1 | - | 3 | 318 | |||
BC106838.2 | - | 3 | 359 | 40044906 |