Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100pbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029036.2 | NP_083312.1 | 5, 6 | 1191 | ||
GenBank mRNA | AK015229.1 | - | 5, 6 | 1191 | ||
AK030636.1 | - | 7, 8 | 885 | |||
AK043160.1 | - | 3, 4 | 195 | |||
AK046756.1 | - | 5, 6 | 2237 | |||
AK146310.1 | - | 5, 6 | 1254 | |||
BC038329.1 | - | 7, 8 | 841 | 1548746 |